Would you like to log in again?
E. coli Exonuclease III
Supplier: New England Biolabs
M0206S M0206L M0206L M0206S
101228-196EA 282 USD InStock
CA101228-196 101228-196 CA101228-198 101228-198
E. coli Exonuclease III
Enzymes
Catalyzes the stepwise removal of mononucleotides from 3´-hydroxyl termini of duplex DNA.
A limited number of nucleotides are removed during each binding event, resulting in coordinated progressive deletions within the population of DNA molecules. The preferred substrates are blunt or recessed 3´-termini, although the enzyme also acts at nicks in duplex DNA to produce single-strand gaps. The enzyme is not active on single-stranded DNA, and thus 3´-protruding termini are resistant to cleavage. The degree of resistance depends on the length of the extension, with extensions 4 bases or longer being essentially resistant to cleavage. This property can be exploited to produce unidirectional deletions from a linear molecule with one resistant (3´-overhang) and one susceptible (blunt or 5´-overhang) terminus. Exonuclease III activity depends partially on helical structure and displays sequence dependence (C>A=T>G). Temperature, salt concentration and the ratio of enzyme to DNA greatly affect enzyme activity, requiring reaction conditions to be tailored to specific applications. Exonuclease III has also been reported to have RNase H, 3´-phosphatase and AP-endonuclease activities.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty