Would you like to log in again?
Msp I Methyltransferase
Supplier: New England Biolabs
M0215S M0215S
101228-234EA 85 USD InStock
CA101228-234 101228-234
Msp I Methyltransferase
Enzymes
MspI Methyltransferase recognizes the same sequence as the HpaII Methyltransferase, but modifies the external cytosine residue (C5) of the sequence CCGG.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty