Would you like to log in again?
dam Methyltransferase
Supplier: New England Biolabs
M0222L M0222S M0222L M0222S
101228-250EA 87 USD InStock
CA101228-250 101228-250 CA101228-248 101228-248
dam Methyltransferase
Enzymes
dam Methyltransferase modifies the adenine residue (N6) of the sequence GATC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty