Would you like to log in again?
Mung bean Nuclease
Supplier: New England Biolabs
M0250L M0250S M0250S M0250L
101228-274EA 302 USD InStock
CA101228-274 CA101228-276 101228-274 101228-276
Mung bean Nuclease
Enzymes
A single-strand specific DNA and RNA endonuclease which will degrade single-stranded extensions from the ends of DNA and RNA molecules, leaving blunt, ligatable ends.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty