Would you like to log in again?
VENT® DNA Polymerase, New England Biolabs
Supplier: New England Biolabs
VENT™
M0254S M0254L M0254S M0254L
101228-290EA 332 USD InStock
CA101228-290 CA101228-292 101228-290 101228-292
VENT® DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VentR DNA Polymerase was the first high-fidelity thermophilic DNA polymerase available for PCR
The fidelity is 5-15-fold higher than  that observed for Taq DNA Polymerase, and is derived in part from an integral 3´→ 5´ proofreading exonuclease activity in Vent DNA Polymerase. Greater than 90% activity remains following a 1 hour incubation at 95°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty