Would you like to log in again?
Uracil Glycosylase Inhibitor
Supplier: New England Biolabs
M0281S M0281S M0281L M0281L
101228-340EA 333 USD InStock
CA101228-340 CA101228-342 101228-340 101228-342
Uracil Glycosylase Inhibitor
Enzymes
The Uracil Glycosylase Inhibitor (UGI) of Bacillus subtilis bacteriophage PBS1 is a small protein (9.5 kDa) which inhibits E.coli uracil-DNA glycosylase (UDG) as well as UDG from other species. Inhibition of UDG occurs by reversible protein binding with a 1:1 UGD:UGI stoichiometry. UGI is capable of dissociating UDG-DNA complexes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty