Would you like to log in again?
Antarctic Phosphatase
Supplier: New England Biolabs
M0289L M0289S M0289L M0289S
101228-348EA 329 USD InStock
101228-348 CA101228-350 101228-350 CA101228-348
Antarctic Phosphatase
Enzymes
Antarctic Phosphatase catalyzes the dephosphorylation of 5´ and 3´ ends of DNA and RNA phosphomonoesters.
Antarctic Phosphatase also hydrolyzes ribo-, as well as deoxyribonucleoside triphosphates (NTPs and dNTPs). Antarctic Phosphatase can be used in many molecular biology applications, such as the removal of phosphorylated ends of DNA and RNA, for subsequent use in cloning or end-labeling of probes. In cloning, dephosphorylation prevents religation of linearized plasmid DNA. The enzyme acts on 5´ protruding, 5´ recessed and blunt ends. Antarctic Phosphatase may also be used to degrade unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing. The enzyme is completely and irreversibly inactivated by heating at 70°C for 5 minutes, thereby making removal of Antarctic Phosphatase prior to ligation or end-labeling unnecessary.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty