Would you like to log in again?
E. coli Exonuclease I
Supplier: New England Biolabs
M0293S M0293L M0293L M0293S
101228-356EA 318 USD InStock
CA101228-356 101228-356 CA101228-358 101228-358
E. coli Exonuclease I
Enzymes
Catalyzes the removal of nucleotides from single-stranded DNA in the 3' to 5' direction.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty