Would you like to log in again?
Thermostable Inorganic Pyrophosphatase
Supplier: New England Biolabs
M0296S M0296S M0296L M0296L
101228-360EA 333 USD InStock
CA101228-360 CA101228-362 101228-360 101228-362
Thermostable Inorganic Pyrophosphatase
Enzymes
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate: P2O7-4 + H2O -> 2HPO4-2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty