Would you like to log in again?
RNase H
Supplier: New England Biolabs
M0297S M0297L M0297S M0297L
101228-364EA 329 USD InStock
CA101228-364 CA101228-366 101228-364 101228-366
RNase H
Enzymes
Ribonuclease H (RNase H) is an endoribonuclease which specifically hydrolyzes the phosphodiester bonds of RNA which is hybridized to DNA
This enzyme does not digest single- or double-stranded DNA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty