Would you like to log in again?
Endonuclease VIII
Supplier: New England Biolabs
M0299S M0299S M0299L M0299L
101228-372EA 347 USD InStock
CA101228-372 101228-372 CA101228-374 101228-374
Endonuclease VIII
Enzymes
Endonuclease VIII from E.coli acts as both an N-glycosylase and an AP-lyase.
The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating an apurinic (AP site). The AP-lyase activity cleaves 3´ and 5´ to the AP site leaving a 5´ phosphate and a 3´ phosphate. Damaged bases recognized and removed by Endonuclease VIII include urea, 5, 6- dihydroxythymine, thymine glycol, 5-hydroxy-5- methylhydanton, uracil glycol, 6-hydroxy-5, 6-dihydrothymine and methyltartronylurea. While Endonuclease VIII is similar to Endonuclease III, Endonuclease VIII has β and δ lyase activity while Endonuclease III has β lyase activity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty