Would you like to log in again?
T7 Endonuclease I, New England Biolabs
Supplier: New England Biolabs
M0302S M0302L M0302S M0302L
101228-380EA 308 USD InStock
CA101228-380 CA101228-382 101228-380 101228-382
T7 Endonuclease I, New England Biolabs
Enzymes
T7 Endonuclease I recognizes and cleaves non-perfectly matched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA and more slowly, nicked double-stranded DNA
The cleavage site is at the first, second or third phosphodiester bond that is 5´ to the mismatch. The protein is the product of T7 gene 3.
Delivery information: Supplied with 10X Reaction Buffer, NEBuffer 2
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty