Would you like to log in again?
Desoxyribonuclease I (DNase I), New England Biolabs
Supplier: New England Biolabs
M0303L M0303S M0303S M0303L
101228-384EA 311 USD InStock
CA101228-384 CA101228-386 101228-384 101228-386
Desoxyribonuclease I (DNase I), New England Biolabs
Enzymes
DNase I, (RNase-free) is an endonuclease that nonspecifically cleaves DNA to release di-, tri- and oligonucleotide products with 5´-phosphorylated and 3´-hydroxylated ends
DNase I acts on single- and double-stranded DNA, chromatin and RNA:DNA hybrids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty