Would you like to log in again?
QUICK-LOAD® DNA Ladder, 1 kb, New England Biolabs
Supplier: New England Biolabs
N0468L N0468S N0468L N0468S
101228-426EA 75 USD InStock
101228-426 CA101449-372 101449-372 CA101228-426
QUICK-LOAD® DNA Ladder, 1 kb, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
Quick-Load® 1 kb DNA Ladder is a premixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye
The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 0.5-10.0 kilobases. The 3.0 kb fragment has increased intensity to serve as reference bands.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty