Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lambda DNA-Hind III Digest, New England Biolabs
Supplier: New England Biolabs
N3012S N3012S N3012L N3012L
101228-436EA 77 USD InStock
101228-436 CA101228-434 CA101228-436 101228-434
Lambda DNA-Hind III Digest, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
The HindIII digest of lambda DNA (cI857ind1 Sam 7) yields 8 fragments suitable for use as molecular weight standards for agarose gel electrophoresis
The approximate mass of DNA in each of the bands is provided (assuming a 1.0 μg load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty