Would you like to log in again?
pUC19 Vector, New England Biolabs
Supplier: New England Biolabs
N3041L N3041L N3041S N3041S
101228-480EA 84 USD InStock
CA101228-480 101228-480 CA101228-478 101228-478
pUC19 Vector, New England Biolabs
Vectors, Plasmids and Libraries
pUC19 is a commonly used plasmid cloning vector in E.coli
The molecule is a small double-stranded circle, 2686 base pairs in length, and has a high copy number. pUC19 carries a 54 base-pair multiple cloning site polylinker that contains unique sites for 13 different hexanucleotide-specific restriction endonucleases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty