Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
DNA Ladder, 1 kb Plus, New England Biolabs
Supplier: New England Biolabs
N3200L N3200S N3200S N3200L
101228-484EA 259 USD InStock
CA101228-484 101228-484 CA101228-486 101228-486
DNA Ladder, 1 kb Plus, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
A number of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis
This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference points. The approximate mass of DNA in each of the bands is provided (assuming a 1.0 μg load) for approximating the mass of DNA in comparably intense samples of similar size. The 2 log dna ladder comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
The NEB 2 log ladder is available in two sizes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty