Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
DNA Ladder, 100 bp, New England Biolabs
Supplier: New England Biolabs
N3231L N3231S N3231S N3231L
101228-490EA 66 USD InStock
CA101228-490 101228-490 CA101228-488 101228-488
DNA Ladder, 100 bp, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
A number of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis.
The digested DNA includes fragments ranging from 100 to 1517 base pairs. The 500 and 1000 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each of the bands is provided (assuming a 0.5 μg load) for approximating the mass of DNA in comparably intense samples of similar size.
Delivery information: Supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty