Would you like to log in again?
Factor Xa Protease, New England Biolabs
Supplier: New England Biolabs
P8010S P8010L P8010L P8010S
101228-574EA 353 USD InStock
CA101228-574 101228-574 CA101228-576 101228-576
Factor Xa Protease, New England Biolabs
Protein Extraction Protein Extraction Reagents
Factor Xa cleaves after the arginine residue in its preferred cleavage site Ile-(Glu or Asp)-Gly-Arg
It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate. The most common secondary site, among those that have been sequenced, is Gly-Arg. There seems to be a correlation between proteins that are unstable in E. coli and those that are cleaved by Factor Xa at secondary sites; this may indicate that these proteins are in a partially unfolded state. Factor Xa will not cleave a site followed by proline or arginine.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty