Would you like to log in again?
Endoproteinase GluC
Supplier: New England Biolabs
P8100S P8100S
101228-582EA 107 USD InStock
CA101228-582 101228-582
Endoproteinase GluC
Enzymes
Endoproteinase GluC (Staphylococcus aureus Protease V8) is a serine proteinase which selectively cleaves peptide bonds C-terminal to glutamic acid residues
Endoproteinase GluC also cleaves at aspartic acid residues at a rate 100-300 times slower than at glutamic acid residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty