Would you like to log in again?
TaqI-v2, New England Biolabs
Supplier: New England Biolabs
R0149L R0149T R0149M R0149T R0149L R0149S R0149M R0149S
101228-810EA 295 USD InStock
101228-810 CA101228-812 101228-812 CA101228-814 CA101228-816 101228-814 101228-816 CA101228-810
TaqI-v2, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that carries a TaqαI overproducing plasmid (F. Barany using an NEB clone). TaqαI has a two amino acid replacement at its amino terminus. This allows for a higher level of expression without interfering with its catalytic properites.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty