Would you like to log in again?
OPTI-MAX INLET CTRG CHK VALVE VARIAN
Supplier: OPTIMIZE TECHNOLOGIES, INC.
Cartridge^ OPTI-MAX* Inlet Cartrodge Check Valve 1/8 inch Ceramic Ball & Seat Stainless Steel Cartridge Varian ProStar 9000 Series 230 240
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
OPTI-MAX INLET CTRG CHK VALVE VARIAN 10123-432 38-38-04790 Each (1Inch) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty