Would you like to log in again?
EXP®2 Stem Trap Kits and Replacement Stems, 5.0 µm HALO®
Supplier: Optimize Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
EXP®2
15-04014-HU 15-04020-HQ 15-04020-HR 15-04008-HS 15-04020-HV 15-04014-HR 15-03992-HU 15-04001-HR 15-04008-HT 15-04020-HU 15-04008-HR 15-04001-HQ 15-03992-HR 15-04008-HQ 15-04020-HS 15-04001-HP 15-04020-HT 15-04014-HV 15-04008-HU 15-04014-HQ 15-04001-HS 15-03992-HP 15-04001-HU 15-03992-HV 15-04001-HV 15-04020-HP 15-04014-HP 15-03992-HS 15-04008-HV 15-04001-HT 15-04014-HT 15-04014-HS 15-04008-HP 15-03992-HT 15-03992-HQ
10123-344PK 1434.22 USD InStock
10123-344 10123-322 10123-464 10123-324 10123-346 10123-466 10123-326 10123-348 10123-468 10123-328 10122-838 10122-836 10123-340 10123-460 10123-342 10123-320 10123-462 10122-842 10122-840 10122-844 10123-318 10123-454 10123-332 10123-456 10123-334 10123-458 10123-336 10123-314 10123-338 10123-316 10123-452 10123-330 10122-830 10122-834 10122-832
EXP®2 Stem Trap Kits and Replacement Stems, 5.0 µm HALO®
Chromatography Columns
The entire EXP®2 stem trap and reusable holder are only slightly larger than a standard HPLC fitting. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports.
  • Rated to 20000+ psi (1400+ bar)
  • Hand-tight and wrench-tight configurationwith auto-adjusting ZDV connection
  • Low-volume/low-dispersion cartridges
  • For nano flow applications
When tightened by hand, the EXP2 stem trap seals to 8700+ psi. All configurations incorporate wrench-flats to enable flawless sealing to 20000+ psi (1400+ bar). The unique packed floating stem installs directly into any 10-32 port and automatically adjusts to provide a perfect ZDV connection. The EXP system enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Specialized features, patented technology, precision engineering and state-of-the-art manufacturing make the new EXP2 stem trap an unbeatable choice for ultra high-pressure nano-flow trapping applications. The new EXP2 design incorporates integral ferrule into the front-end of the holder. No separate ferrule to lose.
Ordering information: Custom packing available.
Delivery information: Kit includes EXP®2 stem holder with fittings and 3 EXP® stem traps (with bed volumes from 0.17 to 2.6 µl), intended for many repeat uses.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty