Would you like to log in again?
Valor® 2000 Compact Bench Scales, Ohaus
Supplier: Ohaus
Valor®
30035684 30035683 30035440 30035441 30035442 30035439 30035682 30035685
10123-882EA 630.85 USD InStock
10123-882 10123-884 10123-886 10123-878 10123-888 10123-890 10123-892 10123-880
Valor® 2000 Compact Bench Scales, Ohaus
Balances and Scales
The Valor® 2000 water-resistant food scale raises the bar and sets a new industry standard with the winning combination of speed and durability
These compact bench scales are easy to clean and easy to transport. They provide a stabilization time of less than 0.5s and feature a power jack auto-closed design. Additional standard features include a level bubble, fixed weighing pan, overload and underload indicators, low battery indicator, AC adapter, and rechargeable battery.
These rapid-response food scales enhance productivity in harsh environments.
Available in both stainless steel and plastic housing designs, the Valor® 2000 offers large LED front and rear displays and puncture-resistant keypads, making it the premiere value scale for use in dry or washdown environments.
Certifications: UL and C-UL listed. CSA certified.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty