Would you like to log in again?
VWR® Volumetric Flasks without Stopper, Amber Glass, Class A, Serialized
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
134.459.08A 131.459.11A 134.459.06A 134.459.04A 131.459.09A 131.459.10A 134.459.02A 134.459.08A 134.459.05A
10124-396CS 1081.83 USD InStock
10124-396 10124-032 10124-394 10124-030 10119-554 10124-028 10124-038 10124-036 10124-034
VWR® Volumetric Flasks without Stopper, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Made from borosilicate 3.3 expansion glass, flasks are serialized and supplied without stopper.
  • Borosilicate glass construction
  • Individually serialized
  • Comes without a stopper
Flasks are made with a graduation mark for accurate measurements.
Ordering information: Flasks come with individual certificate of compliance.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty