Would you like to log in again?
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Amber Glass, Class A, Serialized
Supplier: Avantor
VWR®
131.479.05A 131.479.02A 131.479.07A 131.479.06A 131.479.08A 131.479.01A 131.479.04A 131.479.03A
10124-098CS 628.07 USD InStock
10124-098 10124-096 10124-194 10124-192 10124-106 10124-104 10124-102 10124-100
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with PTFE stopper.
Ordering information: Flasks come with individual certificate of compliance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty