Would you like to log in again?
VWR® Color-Coded Autoclavable Instrument Marking Tape
Supplier: VWR
VWR-IMT-6-C VWR-IMT-3-C VWR-IMT-26-C VWR-IMT-5-C VWR-IMT-37-C VWR-IMT-2-C VWR-IMT-1-C VWR-IMT-4-C VWR-IMT-58-C
10124-154CS 6.97 USD InStock
10124-154 10124-142 10124-152 10124-150 10124-148 10124-158 10124-146 10124-156 10124-144
VWR® Color-Coded Autoclavable Instrument Marking Tape
Tapes
These 9 mm x5m, vinyl, self-stick tapes color code autoclave items.
  • Non-toxic
  • Vinyl
  • Self-stick tapes
Tapes are designed for use on items such as surgical instruments, laboratory glassware, medical waste, and more for easy identification. These tapes attach securely and safely to any clean surface and resist water, oil, and acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty