Would you like to log in again?
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialized
Supplier: VWR
VWR®
131.236.08 131.236.01 131.236.02 131.236.05 131.236.07 131.236.04 131.236.06 131.236.03
10124-074CS 870.52 USD InStock
10124-074 10124-084 10124-072 10124-170 10124-070 10124-168 10124-398 10124-166
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialized
Flasks Volumetric Flasks
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty