Would you like to log in again?
STORAGE BOX/RACK BLUE 12X32 VI
Supplier: MicroSolv Technology Corp
Rack/box^ Storage Box/Rack with cover, Polypropylene, for 12x32mm autosampler vials, blue plastic, 130mmx130mmx45mm. 81 cavities with alpha numeric coding of all 4 sides as well as the cavity bottom.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STORAGE BOX/RACK BLUE 12X32 VI 10124-280 9502SB-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty