Would you like to log in again?
KLEENGUARD® A35 Liquid and Particle Protection Coveralls, Kimberly-Clark
Supplier: Kimberly-Clark
38935 38952 38941 38932 38917 38949 38951 38930 38953 38918 38923 38942 38927 38916 38921 38938 38934 38919 38922 38929 38925 38926 38948 38937 38939 38950 38920 38943
10124-708CS 247.35 USD InStock
10124-708 10124-706 10124-704 10124-726 10124-724 10124-768 10124-770 10124-712 10124-778 10124-776 10124-710 10124-774 10124-730 10124-772 10124-718 10124-716 10124-758 10124-714 10124-780 10124-700 10124-766 10124-722 10124-720 10124-764 10124-784 10124-662 10124-760 10124-782
KLEENGUARD® A35 Liquid and Particle Protection Coveralls, Kimberly-Clark
Disposable Garments
The KLEENGUARD® A35 Liquid & Particle Protection Coveralls are designed for economical liquid and particulate barrier protection.
A white microporous film laminate for use in contact with hazardous particles or liquids, such as liquid handling, pressure washing, maintenance and general manufacturing.
Certifications: ANSI/ISEA 101-1996 sizing standard.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty