Would you like to log in again?
Mega Racks® 10 to 13 mm Tube Storage Racks
Supplier: Heathrow Scientific
HS24322A HS24322E
10124-790CS 226.69 USD InStock
10124-790 10124-742
Mega Racks® 10 to 13 mm Tube Storage Racks
Racks Tube Racks
Fully-assembled, Mega tube racks are for high throughput labs requiring large sample storage.
  • Sturdy and light weight material for easy and secure transport of blood collection tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
Racks are manufactured from polypropylene and are autoclavable.
High-capacity storage for 5 to 7 ml tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty