Would you like to log in again?
Blood Collection Tray
Supplier: Heathrow Scientific
HS120261
10126-318EA 164.3 USD InStock
10126-318
Blood Collection Tray
Trays/Dishes/Pots Blood Collection Trays
Organize phlebotomy, I.V.start or catheter insertion supplies with this spacious, robust and easy to clean carry tray.
  • Lightweight easy-to-clean ABS construction
  • Comfort grip handle tested to hold up to 10 kg
  • Removable handle allows the tray to be stored flat, saving valuable lab space
The lightweight tray features a comfort grip handle and customizable compartments. A 60-place tube rack with 17 mm wells is included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty