Would you like to log in again?
FlexBrush™ Brushes and Covers, Micronova
Supplier: Micronova
FlexBrush™
FBC-1111 FBC-68 BRWM-22512J FBC-611IR FB-12 FBC-6AA FBC-611
10153-664CS 259.79 USD InStock
10153-664 76415-988 68400-718 76214-300 10126-342 89092-120 68400-720
FlexBrush™ Brushes and Covers, Micronova
Brushes
A flexible 12" brush with optional covers in polyester or microfiber. The FlexBrush™ with attached handle is threaded and can be used with a STA series plastic, ACME threaded handle for extended reach.
  • Ideal for cleaning over and around piping and fittings
  • Flexibility of the head allows users to 'shape' the head for custom cleaning applications
  • Available in J or Hook shape
  • Stainless steel or zinc twisted wire core
  • FDA approved materials
Polyester or microfiber covers are lined with 100% knit polyester to easily slide onto the FlexBrush™, containing the bristles. The covers provide the cleaning of a mop with the flexibility of a brush.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty