Would you like to log in again?
Carts and Buckets for Cleanroom Use, Texwipe®
Supplier: Texwipe
TX7046 TX7046E
10127-090EA 1295.49 USD InStock
10127-090 10127-092
Carts and Buckets for Cleanroom Use, Texwipe®
Carts
Stainless steel cart for transporting buckets and bucket systems. Compatible with all stainless steel Texwipe™ buckets.
  • Autoclave safe
  • Two bucket cleaning system
  • Use with cleaning agents, disinfectants, and other solutions in cleanroom environments
  • Compatible with all cleanroom string mops and most wringers
This cart has a protective edge to keep your buckets in place and capture spills.
Can add extender to convert into a 3 bucket system (TX7046E).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty