Would you like to log in again?
Pierce-It™ Closures, Simport
Supplier: Simport
T407-12BK T407-16Y T407-16W T407-12B T407-12L T407-12GY T407-16BK T407-16L T407-12W T407-16G T407-16B T407-16GY T407-16R T407-12R T407-12Y T407-12G
10127-380CS 113.96 USD InStock
10127-380 10127-360 10127-370 10127-362 10127-350 10127-372 10127-364 10127-352 10127-374 10127-366 10127-354 10127-376 10127-368 10127-378 10127-356 10127-358
Pierce-It™ Closures, Simport
Closures Test/Sample Tube Closures
Made of thermoplastic elastomer, these disposable closures produce a firm, leak-resistant seal for glass and plastic tubes
  • The flexible plastic cap can be pierced as often as needed
  • Will not interfere with most common chemistry, coagulation, and drug monitoring methodologies
  • Can be stored in the refrigerator, in the freezer, or at room temperature
  • Withstand agitation in a test tube Vortex mixer
  • Available in 8 colors for easy identification
Two sizes, 13mm and 16mm, fit a variety of tubes including glass evacuated blood drawing tubes. Will also fit 12mm I.D. cuture tubes.
They will protect samples from evaporation and contamination. They can be easily applied and removed with one hand. These closures can also be punctured to allow through-cap sampling via closed-tube instrumentation systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty