Would you like to log in again?
Middlebrook 7H9 Broth
Supplier: Hardy Diagnostics
U32 C32 C62
10127-468PK 135 USD InStock
10127-468 75993-550 76045-420
Middlebrook 7H9 Broth
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of Mycobacterium spp. from sterile body sites.
Hardy Diagnostics Middlebrook 7H9 Broth and Middlebrook 7H9 Broth with Tween® 80 are recommended for use in the cultivation of Mycobacterium spp. from sterile fluids. The media are also used for preparing dilutions of mycobacteria for antimicrobial testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty