Would you like to log in again?
Transport Swabs, Amies Gel with Charcoal, Single Swab
Supplier: Hardy Diagnostics
TransPorter®
4116BX
10127-474PK 165.25 USD InStock
10127-474
Transport Swabs, Amies Gel with Charcoal, Single Swab
Media for Microbiology Dehydrated Media for Microbiology
Suitable for ear, nose, throat, and eye cultures.
  • Regular aluminum wire shaft
  • Rayon mini tip
  • Orange cap
A single sterile soft rayon swab on a straight aluminum wire with a round bottom plastic tube containing 5 ml of modified Amies with charcoal semi-solid transport medium. Charcoal may not be suitable for preparing gram stain slides. Suitable for ear, nose, throat, and eye cultures. Also male urethral samples for Neisseria gonorrhoeae.
The 'Venturi' (hourglass) design locks in the media and specimen, prevents shifting of media, prevents exposure to air, packaged in a clear oxygen barrier wrapper, keeps out oxygen, tamper-evident seal.
Amies media is Stuart's original media formula modified by replacing the glycerophosphate in the original formula for an inorganic phosphate buffer. The metabolism of glycerophosphate by coliform organisms and other Gram-negative rods, in Stuart's original formulation, resulted in proliferation of these organisms from throat, wound and fecal specimens. Amies also found that NaCl at 0.3% w/v was optimal for the preservation of Neisseria gonorrhoeae. Calcium and magnesium salts were added in the belief that these ions were of importance in controlling the permeability of the bacterial cells and contributed to their survival.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty