Would you like to log in again?
Signature Series™ Flex™ Dehydrants, Epredia
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8215 8155 8301R 8101 8115 8215 8301R 8201 8101 8201 8315 8155
89369-384CS 219.3 USD InStock
89369-384 CA82017-788 CA41810-478 82017-788 84000-040 10128-144 10128-142 CA84000-040 41810-478 14214-928 CA14214-928 CA89369-384
Signature Series™ Flex™ Dehydrants, Epredia
Stains Microscopy and staining reagents
Epredia™ Signature Series™ Flex 100, 95 and 80 dehydrants provides gentle dehydration while demonstrtating brillant cytoplasmic staining.
  • Formulation of methyl and isopropyl alcohols
  • Minimizes over-dehydration in smaller specimens
  • Excellent for lipid extraction, allowing for increased paraffin infiltration
  • May be used as a solvent for staining applications
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty