Would you like to log in again?
Cyto-Tek 2500 Cyto-Centrifuge, Sakura
Supplier: Sakura
4300
10128-162EA 19646.65 USD InStock
10128-162
Cyto-Tek 2500 Cyto-Centrifuge, Sakura
Centrifuges Microcentrifuges
Programmable cyto-centrifuge with processing times from 1 second to 99 minutes 59 seconds in 1 second increments; up to 30 programs in memory
Speed adjusts from 500 to 2,500 rpm in 10-rmp increments. Accommodates a range of samples sizes - 1, 6 and 12 mL; up to 12 specimens at a time.
Lockable rotor lid contains aerosols protecting technician from exposure; removable rotor housing is autoclavable.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty