Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

LB Broth (Luria Bertani, Miller), Quality Biological

Supplier: Quality Biological
340-004-131CS 340-004-721 340-004-721EA 340-004-131 340-004-101CS 340-004-101
10128-246CS 409.61 USD InStock
10128-246 10128-244 10128-254 10128-252 10128-250 10128-248
LB Broth (Luria Bertani, Miller), Quality Biological
Media for Microbiology Prepared Media for Microbiology
LB Agar, Miller and LB Broth, Miller are based on LB Medium as described by Miller for the growth and maintenance of E. coli strains used in molecular microbiology procedures. These are nutritionally rich media designed for growth of pure cultures of recombinant strains.

  • 10 g/L of salt formulation
  • Performance Tested

The higher salt concentration (NaCl) in Bertani facilitates better transport and osmotic balance. Based on this information, researcher’s can select the optimal salt concentration for a specific strain in LB Agar, Miller. LB Broth is suitable for propagation of E. Coli and other bacteriological stocks.

Recommendation
This is the best option for LB Broth is a higher concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.

Performance Tested
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
pH: 7.2-7.4

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research Use Only

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty