Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

LB Broth (Lennox), Quality Biological

Supplier: Quality Biological
340-016-101CS 340-016-101
10128-256EA 52.21 USD InStock
10128-256 10128-258
LB Broth (Lennox), Quality Biological
Media for Microbiology Prepared Media for Microbiology
LB Broth, Lennox is a ready-to-use microbial growth medium used for the cultivation of E. coli.

  • Low (5 g/L) salt formulation
  • Performance Tested

LB Broth is a low-salt formulation in comparison to its counterpart LB Broth, Bertani Miller. Lennox is also nutrient rich containing peptides, amino acids, water soluble vitamins and carbohydrates.LB Broth, Lennox is useful for molecular genetic studies, as well as maintenance and propagation of bacteriological stocks.

Recomendation
This is the best option for LB Broth if a lower concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.

UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99

Certifications: Manufactured in an ISO 9001:2015 certifed company in accordance with ISO-14644 and IEST-RP-006.3; meeting standards for ISO Class 8 and ISO Class 7.

Caution: Research Use Only

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty