Would you like to log in again?
YT Medium 2X, Quality Biological
Supplier: Quality Biological
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
340-023-131 340-023-131CS 340-023-101CS 340-023-101
10128-278CS 622.42 USD InStock
10128-278 10128-276 10128-274 10128-272
YT Medium 2X, Quality Biological
Media for Microbiology Prepared Media for Microbiology
YT Medium 2X is an excellent ready-to-use growth medium, nutritionally rich for the growth of recombinant strains of E. coli.
  • Ready to use
E. Coli will thrive in this medium because of the additional amino acids, nucleotide precursors, vitamins, and other essential metabolites the bacteria can utilize to replicate. The presence of sodium chloride in the medium provides a suitable osmotic environment for growth and replication of bacteria.
YT medium 2X is suitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, DNA plasmid production and production of recombinant proteins, and selective cultivation when appropriate antibiotics are added.
YT Medium 2X components such as tryptone and yeast extracts are found in large quantities within this medium; these components will provide the necessary nitrogen and growth factors that the bacteriophage need to replicate more rapidly and efficiently without depleting the host cells of these critical nutrients.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty