Would you like to log in again?
RNA Gel Buffer 10X (MOPS buffer), Quality Biological
Supplier: Quality Biological
351-059-101
10128-502EA 102.12 USD InStock
10128-502
RNA Gel Buffer 10X (MOPS buffer), Quality Biological
Buffers Electrophoresis Buffers Running Buffers
MOPS buffer is used at 1X for the electrophoresis of RNA as the running buffer to separate RNA samples on agarose and formaldehyde-agarose (denaturing) gels. MOPS buffer is also used for Northern Blots -the hybridization of total RNA or mRNA to a membrane.
  • Liquid, 10X concentration
  • Filtered through a 0.22 µm membrane
  • DNase, RNase tested
MOPS buffer is suitable for use in RNA Gel Electrophoresis, Northern Blot Analysis, and when making formaldehyde-agarose gels and associated running buffer for RNA electrophoresis.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty