Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water nuclease-, protease-free, low endotoxin, deionized, sterile, DEPC treated

Supplier: QUALITY BIOLOGICAL, INC.
Synonyms: Dihydrogen oxide

351-068-721 351-068-161 351-068-721EA 351-068-151 351-068-131CS 351-068-131 351-068-491
10128-520EA 961.11 USD InStock
10128-520 10128-518 10128-516 10128-526 10128-514 10128-524 10128-522
Water nuclease-, protease-free, low endotoxin, deionized, sterile, DEPC treated
Water
DEPC Treated Water is Ultra Pure and prepared by reverse osmosis, passed through fine carbon, de-ionized through two resin beds and serially filtered 1 µm positively charged membranes. The water is subsequently treated with 0.1% (v/v) DEPC (Diethylpyrocarbonate) and incubated and heated overnight at 37°C. Finally, DEPC Treated Water is filtered through a 0.1 µm filter.

  • DNase, RNase and Protease tested

DEPC Treated Water can be used to reconstitute nucleic acids and/or proteins. In addition, it can be used as a diluent or solvent for any molecular biology grade reagent. DEPC Treated Water may be usefully employed in RNA Applications (e.g., Preparations of an RNA probe, DNA Applications (e.g., Subcloning in pUC plasmids), and Protein Applications (e.g., Western Blotting).

Harmonization Code: 3822.00.5090
ECCN: EAR9

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Delivery information: Shipping Condition: ambient

Caution: Research Use Only


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty