Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blocking Solution, Quality Biological

Supplier: Quality Biological

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

351-080-101
10128-532EA 88 USD InStock
10128-532
Blocking Solution, Quality Biological
Buffers Electrophoresis Buffers Blocking Buffers
Blocking is a very important step of western blotting, as it prevents antibodies from binding to the membrane nonspecifically. Blocking is often made with 5% BSA or nonfat dried milk to reduce the background. Nonfat dried milk is often preferred as it is inexpensive and widely available. However, milk proteins are not compatible with all detection labels, so care must be taken to choose the appropriate blocking solution.

  • Reduce non-specific binding

Quality Biological's Blocking Solution is prepared from non-fat dried milk in PBS containing sodium azide 0.02% (w/v) as a preservative

It is often a good strategy to incubate the primary antibody with BSA since it is usually needed in higher amounts than the secondary antibody. Putting it in BSA solution allows the antibody to be reused, if the blot does not give good result. BSA blocking solutions are preferred with biotin and AP antibody labels, and antiphosphoprotein antibodies, since milk contains casein, which is itself a phosphoprotein and biotin, thus interfering with the assay results.

Blocking Solution has been used as a reagent to reduce non specific binding of immunological probes in western blotting procedures. Blocking Solution should be used undiluted. Use 0.1 ml of Blocking Solution per cm2 of filter area. A detergent, e.g., Tween 20, may be added to further reduce nonspecific background.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty