Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RNA Gel Loading Solution 10X, Quality Biological

Supplier: Quality Biological

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

351-081-661 351-081-661EA
10128-536EA 106.48 USD InStock
10128-536 10128-534
RNA Gel Loading Solution 10X, Quality Biological
Buffers Electrophoresis Buffers Loading Buffers
RNA Gel Loading Solution, 10X is prepared according to Sambrook, Fritsch & Maniatis (1989)

  • Contains two dyes, bromphenol blue & xylene cyanole FF
  • Filtered through a 0.22 µm membrane
  • RNase tested

The purpose of this reagent is to ensure that the RNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.

RNA Gel Loading Solution is suitable for RNA Gel Electrophoresis and Northern Blot Analysis.

Contents: Bromphenol blue 0.25%, xylene cyanole FF 0.25%, disodium EDTA dehydrate, glycerol 50%, and DEPC treated water

DNase, RNase and Protease Tested.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty