Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protein Running Buffer 10X, Quality Biological

Supplier: Quality Biological

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

351-132-101
10128-560EA 78.53 USD InStock
10128-560
Protein Running Buffer 10X, Quality Biological
Buffers Electrophoresis Buffers Running Buffers
Protein Running Buffer, 10X is used as the electrophoresis buffer during the stacking and resolve process of sodium dodecyl sulfate - polyacrylamide gel electrophoresis (SDS-PAGE), and is the running buffer for TRIS-glycine gel electrophoresis.

  • Contains 1% SDS
  • Filtered through a 0.22 µm membrane
  • DNase, RNase, and Protease tested

Protein running Buffer, 10X is a Tris-Glycine buffer used for sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) of proteins. It is used as both the anode and cathode buffer. Running Buffer, 10X is supplied as 500ml of 10X concentrate that can be diluted to a 1X solution.

This product is ideally suited for protein gel analysis in biochemical, molecular biological, cell biological, and immunological research studies.

Contents: Tris base, glycine, SDS and Molecular Biology Grade Water

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty