Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, Ultrapure for PCR, deionized, sterile

Supplier: QUALITY BIOLOGICAL, INC.
Synonyms: Dihydrogen oxide

351-161-741 351-161-671EA 351-161-671
10128-570PK 247.85 USD InStock
10128-570 10128-568 10128-566
Water, Ultrapure for PCR, deionized, sterile
Water
PCR Water is useful for PCR Assays (i.e rRT-PCR, etc.).

  • DNase, RNase and Protease tested.
  • Free of contaminants of human, mouse and rat DNA.

PCR Qualified Water is functionally tested in a PCR reaction. PCR Qualified Water is highly purified water and it is reverse osmosis deionized, diethylpyrocarbonate (DEPC) treated. This product is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures. Free of detectable contamination of human, mouse and rat DNA.

Endotoxin and Mycoplasma Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Delivery information: Shipping Condition: ambient

Caution: Research Use Only


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty