Synonyms:
Dihydrogen oxide
351-161-741
351-161-671EA
351-161-671
10128-570PK
247.85
USD
InStock
10128-570
10128-568
10128-566
Water, Ultrapure for PCR, deionized, sterile
Water
PCR Water is useful for PCR Assays (i.e rRT-PCR, etc.).
- DNase, RNase and Protease tested.
- Free of contaminants of human, mouse and rat DNA.
PCR Qualified Water is functionally tested in a PCR reaction. PCR Qualified Water is highly purified water and it is reverse osmosis deionized, diethylpyrocarbonate (DEPC) treated. This product is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures. Free of detectable contamination of human, mouse and rat DNA.
Endotoxin and Mycoplasma Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Delivery information: Shipping Condition: ambient
Caution: Research Use Only