Would you like to log in again?
Tris (1 M) Buffer Solution, pH 9.8, Quality Biological
Supplier: Quality Biological
351-176-131
10128-572EA 142.85 USD InStock
10128-572
Tris (1 M) Buffer Solution, pH 9.8, Quality Biological
Buffers Biological Buffers
Suitable for molecular biology, protein chemistry, and biochemical applications.
  • DNase, RNnase and Protease tested.
  • Filtered through a 0.22 µm membrane
Tris Buffer, 1M pH 9.8 is commonly used in SDS-PAGE and electrophoresis applications.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: This product requires special production. Please allow 2 to 4 weeks for delivery.
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty